Learn More
Greiner Bio-One Insect (Drosophila) Breeding Conical Container
Ideally suited for the cultivation of Drosophila melanogaster.
86.43 € - 171.43 €
Specifications
Bottom Shape | Conical |
---|---|
Material | Polystyrene |
Type | Drosophila Breeding Container |
For Use With (Application) | Cultivation of Drosophila melanogaster |
Product Code | Brand | Material | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Capacity (Metric) | Price | Quantity & Availability | ||||
10517381
|
Greiner Bio-One
199101 |
Polystyrene | 16 mL | N/A | |||||
13067683
|
Greiner Bio-One
205101 |
Polystyrene | 28 mL |
171.43 €
Pack of 1500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10160331
![]() |
Greiner Bio-One
217101 |
Polystyrene | 68 mL |
86.43 €
Pack of 605 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Bottom part of container made of either polystyrene or polypropylene
- Compatible with gas-permeable ceaprene stopper made of water-repellent material
Specifications
Conical | |
Polystyrene | |
Drosophila Breeding Container | |
Cultivation of Drosophila melanogaster |