Learn More
Mettler Toledo™ InPro™ 3253i/SG pH Electrodes
Low-maintenance pH electrode supplied complete with factory-filled, prepressurized liquid electrolyte. Mettler Toledo™ InPro™ 3253i/SG pH Electrodes are suited to a wide scope of applications in biotechnology, pharmaceutical and chemical process industries.
12870.00 SEK
Specifications
Connector Types | K8S |
---|---|
Diameter (Metric) | 12 mm |
Type | InPro™ 3253i/SG pH Electrode |
Includes | Solution Ground |
Material | Glass |
Description
- Extended operating life
- Precise measurement values
- Resistance to measurement problems at the diaphragm
- Built-in temperature sensor
Specifications
K8S | |
InPro™ 3253i/SG pH Electrode | |
Glass | |
0°C to 100°C |
12 mm | |
Solution Ground | |
0 to 14 | |
CE Certified |