Learn More
Welch™ Inlet Separator for CRVpro Direct Drive Rotary Vane Vacuum Pump
For use with CRVpro Direct Drive Rotary Vane Vacuum Pump
Brand: Welch™ 320016
3156.03 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Intake separator separates liquid phase aerosols from gas stream, reducing chemical contaminants in pump oil
Specifications
Inlet Separator | |
Separates liquid phase aerosols from gas stream | |
Rotary Vane Pump |