Learn More
Hydrogen peroxide 30%, BAKER ANALYZED™ ACS, J.T. Baker™
Exceeds ACS Specifications
Brand: JT Baker 2192.2500PE
Quantity | 2.5 L |
---|
View more versions of this product
This item is not returnable.
View return policy
Description
- Trace Impurities (in ppm):
- Silver (Ag) max. 0.01
- aluminum (Al) max. 0.05
- Arsenic (As) max. 0.05
- Barium (Ba) max. 0.02
- Beryllium (Be) max. 0.01
- Calcium (Ca) max. 0.5
- Cadmium (Cd) max. 0.02
- Cobalt (Co) max. 0.01
- Chromium (Cr) max. 0.1
- Copper (Cu) max. 0.01
- Iron (Fe) max. 0.1
- Germanium (Ge) max. 0.05
- Potassium (K) max. 0.1
- Lithium (Li) max. 0.01
- Magnesium (Mg) max. 0.1
- Manganese (Mn) max. 0.01
- Molybdenum (Mo) max. 0.02
- Sodium (Na) max. 0.5
- Nickel (Ni) max. 0.05
- Lead (Pb) max. 0.05
- Strontium (Sr) max. 0.01
- Titanium (Ti) max. 0.1
- Thallium (Tl) max. 0.05
- Vanadium (V) max. 0.01
- Zinc (Zn) max. 0.05
- Zirconium (Zr) max. 0.1
Chemical Identifiers
7722-84-1 | |
34.01 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
OO |
H2O2 | |
MFCD00011333 | |
peroxol |
Specifications
7722-84-1 | |
≤0°C | |
H2O2 | |
1.11 g/cm³ (20°C) | |
OO | |
34.01 | |
max. 0.002% |
∼107°C (1013 hPa) | |
30.0 to 32.0% | |
MFCD00011333 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
peroxol | |
Plastic Bottle | |
2.5 L |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.