missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 6M/l Solution, Scharlab
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
- Molecular Formula: HCl
- Molar Mass: 36.46g/mol
- CAS: 7647-01-0
- EIN ECS No.: 231-595-7
- Molarity: 6.0 mol/l
This volumetric solution was checked by means of potentiometric methods using Scharlau's calcium carbonate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).
Specifications
Specifications
CAS | 7647-01-0 |
Composition | 1mL = 0.21876g HCl |
Concentration | 6.0mol/L |
Density | 1.098g/cm3 |
Traceability to NIST | Traceable to NIST |
Normality | 6.0N |
Quantity | 1 L |
Molecular Formula | ClH |
Physical Form | Solution |
MDL Number | MFCD00011324 MFCD00792839 |
Show More |
Safety and Handling

Product Identifier
- Hydrochloric acid, solution 6 mol/l (6 N)
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Hydrogen chloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction