Learn More
Hydrochloric Acid, 0.1M/l Solution, Scharlab
Product Code. 11312485
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Used in analytical chemistry as a laboratory reagent.
  • Molecular Formula: HCl
  • Molar Mass: 36.46g/mol
  • CAS: 7647-01-0
  • EIN ECS No.: 231-595-7
  • Molarity: 0.5 mol/l
This volumetric solution was checked by means of potentiometric methods using Scharlau's calcium carbonate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).
Specifications
CAS 7647-01-0
Composition 1mL = 0.003646g HCl
Concentration 0.1mol/L
Density 1.00g/cm3 
Traceability to NIST Traceable to NIST
Normality 0.1N
Quantity 1 L
Molecular Formula ClH
Physical Form Solution
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol