Learn More
HTR4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21165-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the family of serotonin receptors, which are G protein coupled receptors that stimulate cAMP production in response to serotonin (5-hydroxytryptamine). The gene product is a glycosylated transmembrane protein that functions in both the peripheral and central nervous system to modulate the release of various neurotransmitters. Multiple transcript variants encoding proteins with distinct C-terminal sequences have been described, but the full-length nature of some transcript variants has not been determined.Specifications
HTR4 | |
Polyclonal | |
Unconjugated | |
HTR4 | |
5 HT 4, 5 HT4, 5 HT4R, 5 hydroxytryptamine receptor 4, HTR4, Serotonin receptor 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15562, 3360 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P97288, Q13639 | |
HTR4 | |
HTR4 Fusion Protein Ag15226 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |