Learn More
BINDER™ HEPA Air Filter
HEPA Air Filter on fresh air supply for unit.
Brand:  BINDER™ 8012-2313
18872.64 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17857128
  • 20760.00 SEK / Each
Only null left
Estimated Shipment: 01-09-2025
Description
  • Accessory
  • Filter classification H14 (acc. to. EN 1822-1:2009, >99.995% at 0.3 μm)
  • Not in conjunction with reinforced heating output, reinforced fan
Specifications
Specifications
M 260 E3.1 Drying/Heating Oven
Plastic
HEPA Filter