Learn More
HDAC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12922-3-AP-20UL
 View more versions of this product
Product Code. 16810884
  • 1645.54 SEK / 20µL
Explore more special offers
Description
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human HDAC2.
The protein encoded by this gene is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein shows sequence similarity to the E. coli RNA polymerase-binding protein HepA. Mutations in this gene are a cause of Schimke immunoosseous dysplasia (SIOD), an autosomal recessive disorder with the diagnostic features of spondyloepiphyseal dysplasia, renal dysfunction, and T-cell immunodeficiency.
Specifications
Specifications
HDAC2
Polyclonal
Unconjugated
HDAC2
HD2, HDAC2, histone deacetylase 2, RPD3, YAF1
Rabbit
Antigen Affinity Chromatography
RUO
15182, 3066, 84577
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunoprecipitation
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P70288, Q92769
Hdac2
HDAC2 Fusion Protein Ag3607
20 μL
Primary
Rat, Human, Mouse