Learn More
Sartorius™ Handle
POA - 406.00€
Specifications
Product Type Handle
Quantity 1 Each
View More Specs
Products 4
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10164171
View Documents
Sartorius™
YAW53
50kg Weights
POA
Each
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
10367251
View Documents
Sartorius™
YAW51
10kg Weights
320.00€
Each
Only null left
Estimated Shipment: 07-08-2025
Add to basket
10663922
View Documents
Sartorius™
YAW52
20kg Weights
406.00€
Each
10543382
View Documents
Sartorius™
YAW50
5kg Weights
277.00€
Each
Specifications
Handle
1 Each