Learn More
Graphite rod, 3.8cm (1.5in) dia x 61cm (24in) long, 99% (metals basis), Thermo Scientific Chemicals
2100.00 SEK - 7705.00 SEK
Specifications
Chemical Name or Material | Graphite rod |
---|---|
Name Note | 3.8cm (1.5 in.) dia. x 61cm (24 in.) long |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD00144065 |
Percent Purity | 99% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15406555
|
Thermo Scientific Alfa Aesar
043800.KG |
1 Pc. |
2100.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15416555
|
Thermo Scientific Alfa Aesar
043800.KJ |
5 Pc. |
7705.00 SEK
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Graphite rod | |
(metals basis) | |
99% |
3.8cm (1.5 in.) dia. x 61cm (24 in.) long | |
MFCD00144065 | |
Rod |
Safety and Handling
missing translation for 'einecsNumber' : 231-955-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures