missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNPAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GNPAT |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16863854
|
Proteintech
14931-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16853854
|
Proteintech
14931-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody recognize the 69 kDa GNPAT protein.
GNPAT is a key enzyme located in the peroxisomal membrane which is essential to the synthesis of ether phospholipids. Mutations in GNPAT are associated with rhizomelic chondrodysplasia punctata type 2, which is characterized by rhizomelic shortening of femur and humerus, vertebral disorders, cataract, cutaneous lesions and severe mental retardation.Specifications
GNPAT | |
Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
O15228, P98192, Q9ES71 | |
14712, 8443, 84470 | |
GNPAT Fusion Protein Ag6735 | |
Primary | |
-20°C | |
GNPAT |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DAP AT, DAPAT, DHAP AT, DHAPAT, GNPAT | |
GNPAT | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title