Learn More
Macherey-Nagel™ Glass Flash Chromatography Column
293.00 EUR - 391.00 EUR
Specifications
Material | Glass |
---|---|
Type | Flash Column |
Includes | with Adapter and PTFE Tap |
Column Type | Flash Chromatography Glass Column |
For Use With (Application) | Suited The Separation of Compounds up to Gram Levels No Expensive Equipment Required |
Product Code | Brand | Length (Metric) | Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Style | Price | Quantity & Availability | ||||
10440132
|
Macherey-Nagel™
727400 |
200 mm | I.D.: 20 mm |
293.00 EUR
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
10073131
|
Macherey-Nagel™
727401 |
400 mm | I.D.: 20 mm |
302.00 EUR
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
10659771
|
Macherey-Nagel™
727403 |
400 mm | I.D.: 25 mm |
313.00 EUR
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||
10164801
|
Macherey-Nagel™
727404 |
300 mm | I.D.: 30 mm |
321.00 EUR
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
10450132
|
Macherey-Nagel™
727402 |
200 mm | I.D.: 25 mm |
302.00 EUR
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
10093141
|
Macherey-Nagel™
727405 |
400 mm | I.D.: 30 mm |
330.00 EUR
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
10690152
|
Macherey-Nagel™
727406 |
300 mm | I.D.: 40 mm |
341.00 EUR
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
10246281
|
Macherey-Nagel™
727407 |
450 mm | I.D.: 40 mm |
391.00 EUR
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
Specifications
Glass | |
with Adapter and PTFE Tap | |
Suited The Separation of Compounds up to Gram Levels No Expensive Equipment Required |
Flash Column | |
Flash Chromatography Glass Column |