Learn More
Deltalab™ Glass Cover Slides
For protection of the sample and lens.
Brand:  Deltalab™ D102460
Product Code. 12095912
  • 122.00€ / Each
Only null left
Estimated Shipment: 01-08-2025
Description
  • Clear, pre-washed glass. Thickness: 0.13 – 0.16 mm.
  • Supplied in cases of a hundred or two hundred units (depending on the code).
  • Each case includes a bag of silica salt, and is supplied vacuum in tropical pack. Easy opening, no need for scissors.
  • New improved box which minimize the risk of pack breakage and ensure a better lock.
Specifications
Specifications
Cover Slide
0.15 mm
Protection of the Sample and Lens
Glass
24 x 60 mm
10 x 100