Learn More
Gilson™ PIPETMANTM Diamond Blister Pack Refill Pipette Tips
Autoclavable or gamma-ray sterlized
900.00 SEK - 2535.00 SEK
Specifications
Color | Clear |
---|---|
For Use With (Application) | Pipetting |
Compatibility | PIPETMAN Pipettes |
Product Type | Air Displacement Tips |
Product Line | Pipetman Diamond |
Product Code | Brand | Volume (Metric) | Filtered | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Filtered | Sterility | Price | Quantity & Availability | |||
15247783
![]() |
Gilson™
F172511 |
100 to 1000 μL | Non-filtered | Sterile |
1080.00 SEK
Pack of 960 |
In Stock
Log in to see stock. |
|||
15237783
![]() |
Gilson™
F172510 |
100 to 1000 μL | Non-filtered | Non-sterile |
955.00 SEK
Pack of 960 |
In Stock
Log in to see stock. |
|||
15707627
![]() |
Gilson™
F172313 |
2 to 30 μL | Filtered | Sterile |
2295.00 SEK
Pack of 960 |
In Stock
Log in to see stock. |
|||
15227783
![]() |
Gilson™
F172311 |
2 to 200 μL | Non-filtered | Sterile |
1025.00 SEK
Pack of 960 |
In Stock
Log in to see stock. |
|||
15257783
![]() |
Gilson™
F172513 |
2 to 200 μL | Filtered | Sterile |
2295.00 SEK
Pack of 960 |
In Stock
Log in to see stock. |
|||
15297773
![]() |
Gilson™
F172211 |
0.1 to 20 μL | Non-filtered | Sterile |
1220.00 SEK
Pack of 960 |
In Stock
Log in to see stock. |
|||
15217783
![]() |
Gilson™
F172310 |
2 to 200 μL | Non-filtered | Non-sterile |
900.00 SEK
Pack of 960 |
In Stock
Log in to see stock. |
|||
15207783
![]() |
Gilson™
F172213 |
0.1 to 10 μL | Filtered | Sterile |
2535.00 SEK
Pack of 960 |
In Stock
Log in to see stock. |
|||
15287773
![]() |
Gilson™
F172210 |
0.1 to 20 μL | Non-filtered | Non-sterile |
1080.00 SEK
Pack of 960 |
Estimated Shipment: 30-06-2025
Log in to see stock. |
|||
Description
- Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
- Features graduations at varying volumes allowing for quick-checks when aspirating samples
- Standard or filtered tips
- Validated for use with 384-well tips: F172211G, F172311G and F17511G
Specifications
Clear | |
PIPETMAN Pipettes | |
Pipetman Diamond |
Pipetting | |
Air Displacement Tips | |
Polypropylene |