Learn More
Liebherr™ GGU Series Table-Height Static Freezers
Suitable for under-worktop installation. Liebherr™ GGU Series Table-Height Static Freezers are simple, reliable freezers with evaporator plate shelves, electronic controls, digital display and temperature alarm. Available with shelves only, with wire bins or with solid drawers.
Brand:  Liebherr™ GGU 1550
Product Code. 15716858
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Static plate-evaporator system: Ensures freezing capacity close to the product for very simple and reliable freezing
  • Electronic Controls with Digital Temperature Display: Provides easy-to-use controls without opening the door, with a visual and audible temperature alarm
  • Doors with reversible hinges: Designed for intensive use with ergonomic, easy to grasp handles
  • Efficient refrigeration system: Reduces energy consumption and operating costs with premium-quality components
  • Environmentally friendly refrigerant: Minimal greenhouse effect and no ozone depletion potential with outstanding energy efficiency