Learn More
Liebherr™ GGU Series Table-Height Static Freezers
Suitable for under-worktop installation. Liebherr™ GGU Series Table-Height Static Freezers are simple, reliable freezers with evaporator plate shelves, electronic controls, digital display and temperature alarm. Available with shelves only, with wire bins or with solid drawers.
Brand: Liebherr™ GGU 1550
This item is not returnable.
View return policy
Description
- Static plate-evaporator system: Ensures freezing capacity close to the product for very simple and reliable freezing
- Electronic Controls with Digital Temperature Display: Provides easy-to-use controls without opening the door, with a visual and audible temperature alarm
- Doors with reversible hinges: Designed for intensive use with ergonomic, easy to grasp handles
- Efficient refrigeration system: Reduces energy consumption and operating costs with premium-quality components
- Environmentally friendly refrigerant: Minimal greenhouse effect and no ozone depletion potential with outstanding energy efficiency