Learn More
GBE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20313-1-AP-150UL
This item is not returnable.
View return policy
Description
GBE1 is a glycogen branching enzyme that catalyzes the transfer of alpha-1,4-linked glucosyl units from the outer end of a glycogen chain to an alpha-1,6 position on the same or a neighboring glycogen chain. Branching of the chains is essential to increase the solubility of the glycogen molecule and, consequently, in reducing the osmotic pressure within cells. Highest level of this enzyme are found in liver and muscle. Mutations in this gene are associated with glycogen storage disease IV (also known as Andersen′s disease).Specifications
GBE1 | |
Polyclonal | |
Unconjugated | |
GBE1 | |
Brancher enzyme, GBE, GBE1, Glycogen branching enzyme | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2632, 288333, 74185 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q04446, Q9D6Y9 | |
Gbe1 | |
GBE1 Fusion Protein Ag14154 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |