Learn More
Galectin-3 Mouse anti-Human, CoraLite 488, Clone: 1C1B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60207-100UL
Description
The Galectin-3 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Galectin-3. This antibody recognizes human, mouse, rat antigen. The Galectin-3 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
Galectin-3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
1C1B2 | |
CoraLite 488 | |
100 μL | |
3958 | |
IgG2b |