Learn More
G418 Sulfate Solution, MP Biomedicals
Shop All MP Biomedical Products
Click to view available options
Quantity:
20 mL
50 mL
Unit Size:
20mL
50mL
Specifications
Specifications
Type | G418 Sulfate |
Quantity | 20 mL |
Chemical Name or Material | G418 Sulfate Solution |