Learn More
BUCHI FlashPure Select Flash Chromatography Cartridges: C18
FlashPure Select cartridges are designed for ultimate and unmatched performance.
POA
Specifications
Material | Polypropylene |
---|---|
Type | C18 |
Resolution | 20 to 35 μm |
Column Type | C18 |
For Use With (Application) | Non-Polar Compounds, Ideal for Peptides |
Product Code | Brand | Size (Metric) Tube Rack | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) Tube Rack | Includes | Price | Quantity & Availability | ||||
16330236
|
BUCHI
140000126 |
120 g | 2 Pc. |
POA
Pack of 2 |
N/A | ||||
16390226
|
BUCHI
140000122 |
12 g | 5 Pc. |
POA
Pack of 5 |
N/A | ||||
16300236
|
BUCHI
140000123 |
20 g | 2 Pc. |
POA
Pack of 2 |
N/A | ||||
16340236
|
BUCHI
140000127 |
220 g | 1 Pc. |
POA
Each |
N/A | ||||
16350236
|
BUCHI
140000128 |
330 g | 1 Pc. |
POA
Each |
N/A | ||||
16310236
|
BUCHI
140000124 |
40 g | 2 Pc. |
POA
Pack of 2 |
N/A | ||||
16380226
|
BUCHI
140000121 |
4 g | 5 Pc. |
POA
Pack of 5 |
N/A | ||||
16320236
|
BUCHI
140000125 |
80 g | 2 Pc. |
POA
Pack of 2 |
N/A | ||||
Description
- Spherical particles
- 13-17 μm silica, 20-35 μm C18
- Functionalized silica cartridges can be reused multiple times
- Large selection of cartridge sizes from 4 g - 3,000 g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
Polypropylene | |
20 to 35 μm | |
Non-Polar Compounds, Ideal for Peptides |
C18 | |
C18 | |
Any Flash System |