Learn More
Fisherbrand™ Combs for FB-VE16-1 Vertical Electrophoresis System
Creates the wells in gels for electrophoresis
Brand:  Fisherbrand™ P1-10C
568.56 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11913035
  • 570.00 SEK / Each
Only null left
Estimated Shipment: 30-10-2025
Specifications
Owl P1-CST Multiple Gradient Gel Caster
16 x 14 cm
0.08 cm
7.87 in.
20 cm
5.5 in.
13 cm
1.04 cm
18 cm
Comb
10
183 μL
6.3 in.
16 cm
0.4 in.
14 cm
7.08 in.