Learn More
Thermo Scientific™ FastRuler DNA-ladder, middenbereik, gebruiksklaar, 100-5000 bp
Perform fast sizing and approximate quantification of dsDNA in high throughput gels and conventional agarose gels with Thermo Scientific™ FastRuler™ DNA Ladders.
Merk: Thermo Scientific™ SM1113
Waarschuwingen:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Beschrijving
FastRuler Middle Range DNA Ladder is specifically designed for fast sizing and approximate quantification of double-stranded DNA in 48 well (or 96-well) high throughput gels, as well as in conventional agarose gels. The ladder is a mixture of five blunt-ended chromatography-purified individual DNA fragments that are easily resolved in a short separation distance (10 to 20 mm) after an 8 to 14 minutes run on the appropriate agarose gel.
The ladder is especially useful for electrophoresis of PCR products.
Highlights
- Fast separation (8 to 14 min)
- Short separation distance (10 to 20 mm)
- Sharp bands, easy-to-remember fragment sizes and quantities
- Ready-to-use -premixed with loading dye
- Supplied with loading dye solution for sample DNA
Specificatie
Agarose | |
Middle Range DNA Ladder | |
100 to 5000 bp | |
Sustainable packaging | |
1000 μL |
Bromophenol Blue | |
Yes | |
Agarose Gels | |
FastRuler |