Learn More
Memmert™ Factory Calibration (3 Temperatures) for UF55
Calibration at Memmert factory for three temperature value.
Brand: Memmert™ UF55D00128
This item is not returnable.
View return policy
Description
- For three temperature values: 100°C, 160°C, and 220°C
Specifications
UF55 Oven | |
Measuring point chamber center | |
Certificate for 3 temperatures | |
Standard Works Calibration Certificate |