Learn More
Fisherbrand™ Extreme Case pH Electrode
High-strength glass pH electrode ideal for measuring hydrofluoric acid up to 1 g / ‡.
Brand:  Fisherbrand™ 11736338
1121.98 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11736338
  • 1215.00 SEK / Each
Only null left
Estimated Shipment: 15-10-2025
Add to basket
Description
Description
  • Extreme pH electrode
  • High-strength glass pH electrode
  • Chemical for acid or concentrated base. 
  • Measured in hydrofluoric acid up to 1 g / ‡
Specifications
Specifications
S7
0 to 14
120 mm
Glass
0°C to 80°C
12 mm