Learn More
EXOC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17593-1-AP-150UL
 View more versions of this product
Product Code. 16867524
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.
Specifications
Specifications
EXOC5
Polyclonal
Unconjugated
EXOC5
EXOC5, exocyst complex component 5, HSEC10, PRO1912, SEC10, SEC10L1, SEC10P
Rabbit
Antigen Affinity Chromatography
RUO
105504, 10640, 60627
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00471, P97878, Q3TPX4
Exoc5
EXOC5 Fusion Protein Ag11524
150 μL
Primary
Human, Mouse, Rat