missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediamine tetraacetic acid disodium salt dihydrate, 99.74%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Unit Size:
100g
1kg
250g
500g
Specifications
Specifications
Color | White |
Quantity | 100 g |
Formula Weight | 372.2 |
Percent Purity | 0.9974 |
Physical Form | Powder |
Chemical Name or Material | Ethylenediamine tetraacetic acid disodium salt dihydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction