Learn More
Memmert™ Ethernet Connection Cable for Computer Interface for UF55
Ethernet connection cable for use with Memmert Ovens.
Brand:  Memmert™ UF55E06189
329.11 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17599445
  • 344.00 SEK / Each
Only null left
Estimated Shipment: 26-08-2025
Description
  • 5 m
  • Accessory
Specifications
Specifications
UF55 Oven
USB Connection Cable
5 m