missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, 96% v/v, Pharmpur™, Scharlab
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
Description
- Synonyms: Ethyl alcohol, Methylcarbinol, Spirit, Spirit of wine
- Molecular Formula: C2H5OH
- Molar Mass: 46.07g/mol
- CAS: 64-17-5
- EIN ECS No.: 200-578-6
- Dielectric Constant: 24.3 at 25°C
Reacts explosively with strong oxidizing agents including chromium trioxide, hydrogen peroxide and inorganic acids such as nitric acid. Can react violently with metals such as magnesium, sodium and potassium to produce hydrogen. May damage gaskets, painted surfaces, protective and sealing grease coatings, materials, rubber and some plastics.
Specifications
Specifications
Melting Point | -117°C |
Absorbance | 0.40 (5.0cm, 240nm) |
Acidity | 0.003% max (as CH3COOH) |
Appearance | Clear, Colorless |
Density | 0.81g/cm3 |
Boiling Point | 78°C |
Quantity | 1 L |
Flash Point | 9°C |
Identification | Passes Test |
Ignition Point | 425°C |
Show More |
Safety and Handling

Product Identifier
- Ethanol 96%; extra pure, Pharmpur, Ph Eur, BP
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction