Learn More
Memmert™ Entry Port 23 mm for UF55, Standard Position Left Side Center/Center
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF55F0
1418.57 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17549455
  • 1485.00 SEK / Each
Only null left
Estimated Shipment: 04-09-2025
Description
  • Entry port with 23 mm diameter
  • Flap and stopper closure
  • Standard position left side center/center
Specifications
Specifications
UF55 Oven
Entry Port
23 mm