Learn More
Memmert™ Entry Port 23 mm for UF55, Custom TBC Back Side
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF55F6
2110.83 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17509465
  • 2205.00 SEK / Each
Only null left
Estimated Shipment: 26-08-2025
Description
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
Specifications
UF55 Oven
Entry Port
23 mm