Learn More
Memmert™ Entry Port 23 mm for UF55, Custom Position TBC Left Side
Optional entry port for Memmert Ovens.
Brand: Memmert™ UF55F4
2110.83 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Entry port with 23 mm diameter
- Flap closure
- Custom position TBC in left side
Specifications
UF55 Oven | |
Entry Port | |
23 mm |