Learn More
Memmert™ Entry Port 23 mm for UF160, Standard Position Right Side Center/Center
Optional entry port for Memmert Ovens.
Brand: Memmert™ UF160F2
1418.57 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Entry port with 23 mm diameter
- Flap and stopper closure
- Standard position right side center/center
Specifications
UF160 Oven | |
Entry Port | |
23 mm |