Learn More
American Educational Products Electromagnetic Spectrum Chart
Unique chart showsvastness and variety of the electromagnetic spectrum.
Brand: American Educational Products 4100
This item is not returnable.
View return policy
Description
Includes explanatory instruction sheet. For grades 5+. Dimensions: 34 x 24 in. (85 x 60cm).
- Wavelength, frequency, frequency band names
- Broadcast allocations, black body curves
- Atomic emission spectra, atmospheric absorption bands
- X-ray absorption, gamma ray absorption
- Radioactive isotope and elementary particle emissions
- Binding energies for neutrons
- Narrow band of visible light is highlighted
Specifications
Visual Electromagnetic Spectrum Poster | |
5 to 8 | |
Explanatory instruction sheet | |
Radio and microwave usage, x-ray and gamma ray absorption |