Learn More
EDTA Disodium Salt Solution, 0.1M, Chem-Lab
Product Code. p-8014298
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
Used in electrophoresis and molecular biology applications. EDTA Disodium Salt Solution, 0.1M, Chem-Lab is a chelator of divalent cations for use in TH titrations. Solution contains 37.224g EDTA diNa · 2H2O per liter of water (0.01 M). Sol A = 10°FH/mL or 5.6°DH/mL.
EDTA Disodium Salt Dihydrate is a chelator of divalent cations. It inhibits enzymes, such as metalloproteases, that require divalent cations for activity. It is also used in many enzyme buffers and as an enzyme inactivator at high concentrations.
Chemical Identifiers
CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Specifications
Melting Point 0°C
Color Colorless
Density 1.02g/cm3
pH ±9
Boiling Point 100°C
Odor Odorless
Quantity 1 L
Solubility Information Soluble
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
Formula Weight 372.24g/mol
Concentration or Composition (by Analyte or Components) 0.1mol/L (± 0.0004, 20°C)
Physical Form Liquid