Learn More
Mettler Toledo™ EasyDirect™ pH PC Software
For use with SevenCompact™ Duo S213 pH/Conductivity Meter.
Brand: Mettler Toledo™ 30323214
2803.09 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Capture and store data from any SevenCompact meter
- Directly print or export the data to an open application such as Excel
Specifications
License EasyDirect pH | |
EasyDirect pH Software | |
Capture and store data from any SevenCompact meter |