missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dyclonine hydrochloride, MP Biomedicals™
Shop All MP Biomedical Products
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
Specifications
Specifications
Color | White |
Quantity | 1 g |
Formula Weight | 325.9 |
Physical Form | Powder |
Chemical Name or Material | Dyclonine hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction