Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, GL
For attaching to a wide variety of apparatus.
23.34 SEK - 2065.00 SEK
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
15844628
|
DWK Life Sciences
286251603 |
GL 14 |
2065.00 SEK
Pack of 100 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
|||||
10355301
|
DWK Life Sciences
286258808 |
GL 32 | N/A | ||||||
15981511
|
DWK Life Sciences
286063403 |
GL 45 |
1925.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10778061
|
DWK Life Sciences
286256207 |
GL 25 | N/A | ||||||
13463726
|
DWK Life Sciences
286253201 |
GL 18 |
1210.00 SEK
Pack of 50 |
In Stock
Log in to see stock. |
|||||
Description
The DURAN™ glass threaded tubes with standardised GL thread according to DIN 12216 are used for attachment to glass apparatus. The nominal size refers to the outside diameter measured on the thread. Available in 5 sizes. The available accessories allow a wide range of applications.
- With standardised GL thread according to DIN 12216
- The nominal size refers to the outside diameter measured on the thread (available in 5 sizes).
- Safe and easy sealing and loosening of glass-glass components
- Grease-free working possible
- Glass articles for further processing