Learn More
DWK Life Sciences DURAN™ Microbiology Bottle Unground Wide Neck
Culture bottles with a wide neck for use with rubber stoppers
Brand:  DWK Life Sciences 211814409
 View more versions of this product
Product Code. 11702919
  • 2760.00 SEK / Pack of 10
Only null left
In Stock
Description
The culture bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the preparation of media or culturing of microorganisms. The bottles comply with ISO 4796-2 and feature a wide conical unground neck for use with rubber stoppers. Suitable for autoclaving at 121 or 134°C or dry heat sterilization.
  • Wide neck for rubber stoppers
  • Wide neck for easier filling and cleaning
  • Retrace code
  • Fired-on, highly durable white print
  • Autoclavable at 121° C or 134°C
  • Supplied without stopper
TRUSTED_SUSTAINABILITY
Specifications
Specifications
500 mL
Borosilicate Glass (3.3)
Autoclavable
Autoclaving of media and reagents; Preparation of media and reagents; Culturing vessel
10/Pk.
86 mm