missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTWD2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24873-1-AP-20UL
This item is not returnable.
View return policy
Description
DTWD2 is a protein coding gene.Specifications
DTWD2 | |
Polyclonal | |
Unconjugated | |
DTWD2 | |
DTW domain containing 2, DTWD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
285605 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8NBA8 | |
DTWD2 | |
DTWD2 Fusion Protein Ag21665 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction