Learn More
Sampling Systems™ Double Bagged - USP Class VI Sterile Scoops
The SteriWare™ Ultra range of Double Bagged Scoops are made from high quality USP class VI HDPE. This range is ideal for demanding applications such as vaccine manufacture, GMP and cleanroom manufacturing.
€ 98.70 - € 156.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17554706
![]() |
Sampling Systems™
9286H-25S |
25 mL |
€ 119.00
Pack of 50 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
|||||
17574706
![]() |
Sampling Systems™
9286H-50S |
50 mL |
€ 122.00
Pack of 50 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
|||||
17524706
![]() |
Sampling Systems™
9286H-100S |
100 mL |
€ 133.00
Pack of 50 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
|||||
17534706
![]() |
Sampling Systems™
9286H-150S |
150 mL |
€ 137.00
Pack of 50 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
|||||
17544706
![]() |
Sampling Systems™
9286H-250S |
250 mL |
€ 98.70
Pack of 25 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
|||||
17564706
![]() |
Sampling Systems™
9286H-500S |
500 mL |
€ 101.00
Pack of 20 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
|||||
17514706
![]() |
Sampling Systems™
9286H-1000S |
1000 mL |
€ 156.00
Pack of 20 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
|||||
Description
- Our range of USP class VI scoops are available in a range of sizes from 25 mL to 1000 mL.
- This premium scoop range is double bagged.
- A box liner is also used giving a third layer of protection.
- It is ideal for demanding applications such as vaccine manufacture, GMP and cleanroom manufacturing.
- The scoops all have an ergonomic handle and are easy to use.
- The volumetric shape means specific volumes can be quickly measured.
- The flat base means they also sit flat which makes them ideal for use with weighing scales and decanting powder.
- The ends of the scoop are tapered in so it makes them easy to empty.
- Made from food grade, FDA and EC compliant HDPE.
- HDPE is more chemically resistant than traditional PS scoops.
- Scoops comply with SAL 10-6.
- High quality medical grade polymer that has also been independently tested to USP 661.
- The scoops are entirely made in cleanroom conditions with final packing carried out in a class 10,000 cleanroom.
- Each scoop is 100% inspected before packing
Specifications
HDPE | |
Sterile |