Learn More
Karl Hecht™ Assistent™ Digital Interval Timers
Excellent for various timing purposes in the laboratory. Karl Hecht™ Assistent™ Digital Interval Timers are programmable up to 99 minutes and 59 seconds.
30.00 € - 32.85 €
Specifications
Display Digital
View More Specs
Products 2
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
10247381
View Documents
Karl Hecht™
41977060
84 x 77 x 14 mm
30.00 €
Each
Only null left
Estimated Shipment: 09-07-2025
Add to basket
10619051
View Documents
Karl Hecht™
41977050
76 x 66 x 18 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • With magnetic back
  • Includes battery
  • Counts down in seconds
  • Memory recall function
  • Individual setting for minutes/seconds
  • LCD display, 4 digits showing minutes/seconds
  • Alarm sounds for 30 seconds when countdown reaches zero
Specifications
Specifications
Digital