missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, ACS Reagent, contains BHT as inhibitor, anhydrous, ≥99.0%, Honeywell Riedel-de Haën™

Product Code. 15674310
Click to view available options
Quantity:
100 mL
250 mL
1 L
6 x 1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent, contains BHT as inhibitor, anhydrous,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C (literature)
CAS Max % ≤100.0000%
Density 0.706 g/mL at 25 °C(lit.)
Boiling Point 34°C to 35°C
Quantity 250 mL
Flash Point −45°C
Packaging Glass Bottle
Physical Form anhydrous
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Formula Weight 74.12
Percent Purity >99%
Grade ACS Reagent
Assay (GC)
Chemical Name or Material Diethyl ether, ACS Reagent, contains BHT as inhibitor, anhydrous, ≥99.0%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.