missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodimethylsilane, 99+%, Thermo Scientific Chemicals

Product Code. 11965791
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 75-78-5 | C2H6Cl2Si | 129.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-78-5
Molecular Formula C2H6Cl2Si
Molecular Weight (g/mol) 129.06
InChI Key LIKFHECYJZWXFJ-UHFFFAOYSA-N
Synonym dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
PubChem CID 6398
IUPAC Name dichloro(dimethyl)silane
SMILES C[Si](C)(Cl)Cl

Specifications

Melting Point -76.0°C
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 70.0°C
Flash Point −9°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2SiCl2
Refractive Index 1.4030 to 1.4050
Quantity 2.5 L
Fieser 03,114; 04,183; 17,113
Specific Gravity 1.06
Solubility Information Solubility in water: reacts. Other solubilities: soluble in proton-free solvents (care must be,taken with ketones and esters),soluble in benzene and ether,decomposes in ethanol
Viscosity 0.6 mPa.s (25°C)
Formula Weight 129.06
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dichlorodimethylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichlorodimethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.