Learn More
Fisherbrand™ Device Cover for Elmasonic X-tra ST Ultrasonic Cleaners
Provides a secure closure for ultrasonic cleaning baths.
1010.00 SEK - 2665.00 SEK
Specifications
Type | Lid |
---|---|
For Use With (Application) | Closure for X-tra ST Ultrasonic Cleaners |
Includes | Handle |
Shape | Flat |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15636084
|
Fisherbrand™
1076807 |
Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H |
2665.00 SEK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||||
15646084
|
Fisherbrand™
1076813 |
Ultrasonic Cleaner, Elmastonic Xtra ST 300 H/500 H |
1010.00 SEK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||||
15656084
|
Fisherbrand™
1076814 |
Ultrasonic Cleaner, Elmasonic Xtra ST 600 H/800 H |
1055.00 SEK
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
|||||
15666084
|
Fisherbrand™
1076810 |
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H |
2345.00 SEK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||||
Description
- Flat design with handle
- Available in a range of sizes for all X-tra ST models
- Manufactured from stainless steel
- Accessory item
Specifications
Lid | |
Closure for X-tra ST Ultrasonic Cleaners | |
Handle | |
Flat |