missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCLK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21699-1-AP-150UL
This item is not returnable.
View return policy
Description
Eukaryotic translation initiation factor 4 gamma 2 (elF4G or DAP5) plays a role in the switch from cap-dependent to IRES-mediated translation during mitosis, apoptosis, and viral infections. eIF4G2 is a ubiquitously expressed member of the translation initiation factor eIF4G family that lacks the EIF4E binding site.Specifications
DCLK1 | |
Polyclonal | |
Unconjugated | |
DCLK1 | |
DCAMKL1, DCDC3A, DCLK, DCLK1, doublecortin like kinase 1, KIAA0369 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13175, 83825, 9201 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08875, O15075, Q9JLM8 | |
DCLK1 | |
DCLK1 Fusion Protein Ag16292 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction