Learn More
CYP21A2 Mouse anti-Human, Unconjugated, Clone: 1G2G6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67421-1-IG-20UL
This item is not returnable.
View return policy
Description
The CYP21A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP21A2. This antibody recognizes human, pig antigen. The CYP21A2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CYP21A2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Pig |
Western Blot, ELISA | |
1G2G6 | |
Unconjugated | |
20 μL | |
1589 | |
IgG1 |