Learn More
Cole-Parmer™ CPC™ Quick Disconnect Two-Cycle Sanitary Couplings
Innovative product design allows for a one-step process for sterilization and media transfer while eliminating dead spots, reducing the number of connections, and eliminating the need for a laminar flow hood
Brand:  Cole-Parmer™ STC2020000
Product Code. 15457383
  • 5660.00 SEK / Each
Only null left
Estimated Shipment: 21-10-2025
Description
  • Offer the additional benefit of both steam on and steam off functionality.
  • Valve can return to the steam position to allow a second steam cycle after media transfer.
  • Terminations on these connectors are set to industry standards for bioreactors and are available in popular sizes of flexible tubing.
  • Wetted surfaces meet requirements for USP Class VI physicochemical tests.
  • All units are made of animal-free polysulfone resin.
Specifications
Specifications
Sanitary x Barb
3/4 in.
Gamma or steam-in-place (SIP)
Polysulfone
3/4 in.