Learn More
Karl Hecht™ Assistent™ Circular Cover Glasses for Microscopy
Preserve and protect specimens on slides. Karl Hecht™ Assistent™ Circular Cover Glasses for Microscopy are manufactured from a super-transparent borosilicate glass, preventing virtually any color distortion.
Brand:  Karl Hecht™ 1001/0020
 View more versions of this product
Product Code. 10620962
Description
Description
  • 0.13 - 0.16mm thickness
  • Excellent flatness
  • With CE marking
  • Optimum resistance to chemical attack
  • Refractive index finely adapted to microscopes
  • High spectral transmission
  • Excellent internal glass quality with very low levels of inclusions, striae, bubbles, streaks, etc.
  • In boxes of 100 pieces, 10 boxes in a carton
Specifications
Specifications
Coverslip
Glass
20 mm
100/Pk.
Circle
#1 (0.13 to 0.16 mm)
CE