Learn More
Marienfeld Superior™ Counting Chamber with Dark Line Double Net Ruling
Marienfeld Superior™ Counting Chamber with double net ruling are precise measuring instruments for investigation of cell suspensions
Brand: Marienfeld Superior™ 06 402 10
This item is not returnable.
View return policy
Description
- Precise measuring instrument for determination of the number of particles in liquids
- Made of special optical glass
- Equipped with two grids
- With 2 haemacytometer cover glasses 20x26 mm
- Individually packed in a transparent plastic box, 10 boxes in a carton
Specifications
Counting chamber, Dark line | |
CE-marked (IVD) | |
Special optical glass | |
0.1 mm |
Burker | |
0.0025 mm2 | |
1 | |
Two cover glasses 20 x 26 mm |