Learn More
Coulter™ Flow-Count™ Fluorospheres
Used to determine absolute counts on the flow cytometer
Brand: Coulter™ 7547053
This item is not returnable.
View return policy
Specifications
IVD, CE, FCM | |
525 to 700 nm | |
Polystyrene | |
30 Days |
2° to 8°C | |
20 mL Flow-Count™ fluorosphere | |
20 mL | |
Fluorescent Polystyrene Beads |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.